Stampa…

LW6 ≥97.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: CAY10585, 3-[[[4-(Adamantan-1-yl)phenoxy]acetyl]amino]-4-hydroxybenzoic acid methyll ester, Methyl 3-(2-(4-(adamantan-1-yl)phenoxy)acetamido)-4-hydroxybenzoate, Methyl 3-[[[4-(adamantan-1-yl)phenoxy]acetyl]amino]-4-hydroxybenzoate

L0334-100MG L0334-25MG
TCIAL0334-100MGEA 517 EUR
TCIAL0334-100MG TCIAL0334-25MG
LW6 ≥97.0% (by HPLC)
LW6

This product is unavailable in the U.S.

Formula: C₂₆H₂₉NO₅
Peso molecolare: 435,51 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD09907563
CAS Number: 934593-90-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 97.0 area%
Elemental analysis (Nitrogen) 2.80 to 3.60 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR