Stampa…

Lycorine hydrochloride monohydrate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Pericolo

L0319-100MG L0319-25MG
TCIAL0319-100MGEA 313 EUR
TCIAL0319-100MG TCIAL0319-25MG
Lycorine hydrochloride monohydrate ≥98.0% (by HPLC, titration analysis)
Lycorine hydrochloride monohydrate
Formula: C₁₆H₁₇NO₄·HCl·H₂O
Punto di fusione: 215°C
Temperatura di stoccaggio: Refrigerata
CAS Number: 6150-58-9
UN: 2811
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +41.0 to +45.0 deg (C=2.9, water)
Water max. 7.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR