Stampa…

Piombo (II) cloruro for perovskite precursor, purified by sublimation

Fornitore: TCI
Pericolo

Synonyms: Lead dichloride

L0291-1G L0291-5G
TCIAL0291-1GEA 55 EUR
TCIAL0291-1G TCIAL0291-5G
Piombo (II) cloruro for perovskite precursor, purified by sublimation
Piombo (II) cloruro
Formula: Cl₂Pb
Punto di ebollizione: 951°C
Punto di fusione: 501°C
MDL Number: MFCD00011157
CAS Number: 7758-95-4
EINECS: 231-845-5
UN: 2291
ADR: 6.1,III
Merck Index: 14,05404
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bde4b379a8597ae2da9181ba4e03b3aebce58873&vid=bde4b379a8597ae2da9181ba4e03b3aebce58873&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystaline


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR