Loganin ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
(1S,4aS,6S,7R,7aS)-Methyl 6-hydroxy-7-methyl-1-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-1,4a,5,6,7,7a-hexahydrocyclopenta[c]pyran-4-carboxylate
L0268-10MG
L0268-50MG
TCIAL0268-10MGEA
75
EUR
TCIAL0268-10MG
TCIAL0268-50MG
Loganin ≥98.0% (by HPLC)
Loganin
Formula:
C₁₇H₂₆O₁₀ Peso molecolare: 390,39 g/mol |
MDL Number:
MFCD00075645 CAS Number: 18524-94-2 EINECS: 242-398-0 Merck Index: 14,05560 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 223.0 to 227.0 deg-C |
Specific rotation [a]20/D | -84.0 to -87.0 deg (C=0.1, MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...