Stampa…

L-tert-Leucine methyl ester ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (S)-Methyl 2-amino-3,3-dimethylbutanoate, (S)-2-Amino-3,3-dimethylbutyric Acid Methyl Ester, H-Tle-OMe, Methyl (S)-2-Amino-3,3-dimethylbutyrate

L0255-200MG L0255-1G
TCIAL0255-200MGEA 84 EUR
TCIAL0255-200MG TCIAL0255-1G
L-tert-Leucine methyl ester ≥98.0% (by GC, titration analysis)
L-tert-Leucine methyl ester
Formula: C₇H₁₅NO₂
Peso molecolare: 145,20 g/mol
Punto di ebollizione: 69°C
MDL Number: MFCD00137409
CAS Number: 63038-26-6
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09ce67e793ff92b14e96451ea47cb16fed88a917&vid=09ce67e793ff92b14e96451ea47cb16fed88a917&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +65.0 to +70.0 deg (C=1、EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR