Stampa…

Lomustine ≥98.0% (by HPLC)

Fornitore: TCI
Pericolo

L0251-50MG L0251-200MG
TCIAL0251-50MGEA 31 EUR
TCIAL0251-50MG TCIAL0251-200MG
Lomustine ≥98.0% (by HPLC)
Lomustine
Formula: C₉H₁₆ClN₃O₂
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00012392
CAS Number: 13010-47-4
EINECS: 235-859-2
UN: 2811
ADR: 6.1,III
Merck Index: 14,05564
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb453ad7180f4f451fd78e9e5830877094d209bc&vid=eb453ad7180f4f451fd78e9e5830877094d209bc&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 89.0 to 93.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR