Stampa…

Leflunomide ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 5-Methyl-N-(4-(trifluoromethyl)phenyl)isoxazole-4-carboxamide, 5-Methyl-N-[4-(trifluoromethyl)phenyl]isoxazole-4-carboxamide

L0250-200MG L0250-1G
TCIAL0250-200MGEA 51 EUR
TCIAL0250-200MG TCIAL0250-1G
Leflunomide ≥98.0% (by HPLC, total nitrogen)
Leflunomide
Formula: C₁₂H₉F₃N₂O₂
Peso molecolare: 270,21 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00867593
CAS Number: 75706-12-6
UN: 2811
ADR: 6.1,III
Merck Index: 14,05432
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be153f844481a0beb0d5a99da5964774a7aa5b41&vid=be153f844481a0beb0d5a99da5964774a7aa5b41&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 166.0 to 170.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR