Lornoxicam ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
6-Chloro-4-hydroxy-2-methyl-N-(2-pyridyl)-2H-thieno[2,3-e][1,2]thiazine-3-carboxamide 1,1-Dioxide, 6-Chloro-4-hydroxy-2-methyl-N-(pyridin-2-yl)-2H-thieno[2,3-e][1,2]thiazine-3-carboxamide 1,1-dioxide
L0239-1G
L0239-5G
TCIAL0239-1GEA
69
EUR
TCIAL0239-1G
TCIAL0239-5G
Lornoxicam ≥98.0% (by HPLC, total nitrogen)
Lornoxicam
Formula:
C₁₃H₁₀ClN₃O₄S₂ Peso molecolare: 371,81 g/mol Punto di fusione: 230°C Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00866163 CAS Number: 70374-39-9 UN: 2811 ADR: 6.1,II Merck Index: 14,05582 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...