Stampa…

Levamisole hydrochloride ≥99.0% (by titrimetric analysis) for biochemical research

Fornitore: TCI EUROPE
Pericolo

Synonyms: (-)-Tetramisole Hydrochloride, (-)-2,3,5,6-Tetrahydro-6-phenylimidazo[2,1-b]thiazole Hydrochloride, (S)-6-Phenyl-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole hydrochloride

L0231-1G L0231-5G
TCIAL0231-1GEA 30 EUR
TCIAL0231-1G TCIAL0231-5G
Levamisole hydrochloride ≥99.0% (by titrimetric analysis) for biochemical research
Levamisole hydrochloride
Formula: C₁₁H₁₂N₂S·HCl
Peso molecolare: 240,75 g/mol
MDL Number: MFCD00012675
CAS Number: 16595-80-5
EINECS: 240-654-6
UN: 2811
ADR: 6.1,III
Merck Index: 14,05459

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 226.0 to 234.0 deg-C
Specific rotation [a]20/D -122.0 to -130.0 deg (C=1, H2O)
Phosphatase inhibition test to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR