Stampa…

Loratadine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Ethyl 4-(8-chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)piperidine-1-carboxylate

L0223-1G L0223-100MG
TCIAL0223-1GEA 65 EUR
TCIAL0223-1G TCIAL0223-100MG
Loratadine ≥98.0% (by HPLC, titration analysis)
Loratadine
Formula: C₂₂H₂₃ClN₂O₂
Peso molecolare: 382,88 g/mol
MDL Number: MFCD00672869
CAS Number: 79794-75-5
Merck Index: 14,05578

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 135.0 to 139.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR