Stampa…

D-Leucine methyl ester hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Fornitore: TCI EUROPE

Synonyms: H-D-Leu-OMe.HCl

L0198-1G L0198-5G
TCIAL0198-1GEA 28 EUR
TCIAL0198-1G TCIAL0198-5G
D-Leucine methyl ester hydrochloride ≥98.0% (by total nitrogen and titration analysis)
D-Leucine methyl ester hydrochloride
Formula: C₇H₁₅NO₂
Peso molecolare: 181,66 g/mol
Punto di fusione: 151°C
MDL Number: MFCD00066115
CAS Number: 5845-53-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D -12.0 to -16.0 deg (C=2, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR