Stampa…

L-Leucine benzyl ester-p-toluenesulphonate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: H-Leu-OBzl.Tos-OH, L-Leucine benzyl ester p-toluenesulphonate salt, H-Leu-OBzl·TosOH

L0195-25G L0195-5G
TCIAL0195-25GEA 96 EUR
TCIAL0195-25G TCIAL0195-5G
L-Leucine benzyl ester-p-toluenesulphonate ≥98.0% (by HPLC, titration analysis)
L-Leucine benzyl ester-p-toluenesulphonate
Formula: C₁₃H₁₉NO₂
Peso molecolare: 393,50 g/mol
MDL Number: MFCD00066117
CAS Number: 1738-77-8
EINECS: 217-095-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 % (total of Ion-Pair)
Purity (Neutralization titration) min. 98.0 %
Melting point 156.0 to 160.0 deg-C
Specific rotation [a]20/D +3.0 to +6.0 deg (C=2, DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR