Stampa…

2-Naphthyl laurate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Lauric Acid 2-Naphthyl Ester

L0122-25G L0122-5G
TCIAL0122-25GEA 150 EUR
TCIAL0122-25G TCIAL0122-5G
2-Naphthyl laurate ≥98.0% (by GC)
2-Naphthyl laurate
Formula: C₂₂H₃₀O₂
Peso molecolare: 326,48 g/mol
MDL Number: MFCD00021608
CAS Number: 6343-73-3
EINECS: 228-741-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 59.0 to 63.0 deg-C
Solubility in Toluene very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR