Stampa…

DL-Leucic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Hydroxy-4-methylpentanoic acid, Leucic acid, DL-2-hydroxy-4-methylvaleric acid, 2-Hydroxy-4-methylvaleric Acid

L0104-25G L0104-500G
TCIAL0104-25GEA 57 EUR
TCIAL0104-25G TCIAL0104-500G
DL-Leucic acid ≥98.0% (by GC, titration analysis)
DL-Leucic acid
Formula: C₆H₁₂O₃
Peso molecolare: 132,16 g/mol
MDL Number: MFCD00004246
CAS Number: 498-36-2
EINECS: 207-860-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 74.0 to 78.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR