Stampa…

2,6-Lutidine-N-oxide ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,6-Dimethylpyridine N-Oxide

L0069-25G L0069-5G
TCIAL0069-25GEA 162 EUR
TCIAL0069-25G TCIAL0069-5G
2,6-Lutidine-N-oxide ≥98.0% (by GC)
2,6-Lutidine-N-oxide
Formula: C₇H₉NO
Peso molecolare: 123,16 g/mol
Punto di ebollizione: 145°C
MDL Number: MFCD00039715
CAS Number: 1073-23-0
EINECS: 214-025-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=748ab20847e1964ef36f13e0015c81f4a22f73a5&vid=748ab20847e1964ef36f13e0015c81f4a22f73a5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR