DL-Leucylglycine ≥98.0% (by titrimetric analysis)
Fornitore: TCI
Formula:
C₈H₁₆N₂O₃ |
MDL Number:
MFCD00021729 CAS Number: 615-82-7 EINECS: 210-449-6 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...