Stampa…

SKLB1002 ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-[(6,7-Dimethoxyquinazolin-4-yl)thio]-5-methyl-1,3,4-thiadiazole, 2-((6,7-Dimethoxyquinazolin-4-yl)thio)-5-methyl-1,3,4-thiadiazole, 6,7-Dimethoxy-4-[(5-methyl-1,3,4-thiadiazol-2-yl)thio]quinazoline

K0060-25MG K0060-100MG
TCIAK0060-25MGEA 149 EUR
TCIAK0060-25MG TCIAK0060-100MG
SKLB1002 ≥98.0% (by HPLC)
SKLB1002
Formula: C₁₃H₁₂N₄O₂S₂
Peso molecolare: 320,39 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD27938707
CAS Number: 1225451-84-2
UN: 2811
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 191.0 to 195.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR