Stampa…

Kenpaullone ≥97.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 9-Bromo-7,12-dihydroindolo[3,2-d][1]benzazepin-6(5H)-one, 9-Bromo-7,12-dihydrobenzo[2,3]azepino[4,5-b]indol-6(5H)-one

K0052-10MG K0052-50MG
TCIAK0052-10MGEA 109 EUR
TCIAK0052-10MG TCIAK0052-50MG
Kenpaullone ≥97.0% (by HPLC)
Kenpaullone
Formula: C₁₆H₁₁BrN₂O
Peso molecolare: 327,18 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD02683595
CAS Number: 142273-20-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Elemental analysis (Nitrogen) 8.00 to 9.00 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR