Stampa…

Ketanserin ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-[2-[4-(4-Fluorobenzoyl)-1-piperidinyl]ethyl]-2,4(1H,3H)-quinazolinedione, 3-(2-(4-(4-Fluorobenzoyl)piperidin-1-yl)ethyl)quinazoline-2,4(1H,3H)-dione

K0051-200MG K0051-1G
TCIAK0051-200MGEA 36 EUR
TCIAK0051-200MG TCIAK0051-1G
Ketanserin ≥98.0% (by HPLC, titration analysis)
Ketanserin
Formula: C₂₂H₂₂FN₃O₃
Peso molecolare: 395,43 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00083392
CAS Number: 74050-98-9
EINECS: 277-680-2
Merck Index: 14,05296

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 231.0 to 235.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR