Stampa…

Ketotifen fumarate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 4-(1-Methylpiperidin-4-ylidene)-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-10(9H)-one fumarate

K0048-1G K0048-5G
TCIAK0048-1GEA 89 EUR
TCIAK0048-1G TCIAK0048-5G
Ketotifen fumarate ≥98.0% (by HPLC, titration analysis)
Ketotifen fumarate
Formula: C₁₉H₁₉NOS
Peso molecolare: 425.50 g/mol
Punto di fusione: 192°C
MDL Number: MFCD00079394
CAS Number: 34580-14-8
EINECS: 252-100-0
Merck Index: 14,05307

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR