Stampa…

Khellin ≥95.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Visammin, 4,9-Dimethoxy-7-methyl-5H-furo[3,2-g]chromen-5-one, 4,9-Dimethoxy-7-methyl-5H-furo[3,2-g][1]benzopyran-5-one

K0039-25G K0039-5G K0039-1G
TCIAK0039-25GEA 222 EUR
TCIAK0039-25G TCIAK0039-5G TCIAK0039-1G
Khellin ≥95.0% (by GC)
Khellin
Formula: C₁₄H₁₂O₅
Peso molecolare: 260,25 g/mol
MDL Number: MFCD00005007
CAS Number: 82-02-0
EINECS: 201-392-8
UN: 2811
ADR: 6.1,III
Merck Index: 14,05309

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 150.0 to 154.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR