Stampa…

5-Oxohexanoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Glurate, 5-Ketohexanoic Acid, 4-Acetylbutyric Acid

K0014-25G K0014-5G
TCIAK0014-25GEA 509 EUR
TCIAK0014-25G TCIAK0014-5G
5-Oxohexanoic acid ≥98.0% (by GC, titration analysis)
5-Oxohexanoic acid
Formula: C₆H₁₀O₃
Peso molecolare: 130,14 g/mol
Punto di ebollizione: 160°C
MDL Number: MFCD00004412
CAS Number: 3128-06-1
EINECS: 221-511-7

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR