Tetrakis(decyl)ammonium bromide ≥98.0% (by titrimetric analysis) per cromatografia
Fornitore: TCI
|
Attenzione
|
Synonyms:
Tetrakis(decyl)ammonium bromide, IPC-TDA-Br, Tetra(decyl)ammonium bromide
Formula:
C₄₀H₈₄BrN |
MDL Number:
MFCD00043166 CAS Number: 14937-42-9 EINECS: 239-013-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Absorbance (230nm) | max. 0.050 |
Absorbance (254nm) | max. 0.040 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...