2-Methylpropanohydrazide ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
I1105-5G
I1105-25G
TCIAI1105-5GEA
131
EUR
TCIAI1105-5G
TCIAI1105-25G
2-Methylpropanohydrazide ≥98.0% (by GC)
2-Methylpropanohydrazide
Formula:
C₄H₁₀N₂O Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00025122 CAS Number: 3619-17-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Bromination Method) | min. 97.0 % |
Melting point | 98.0 to 102.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...