Isodecyl acrylate (mixture of branched chain isomers) stabilizzato
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Acrylic acid Isodecyl ester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
I1104-25G
I1104-100G
TCIAI1104-25GEA
30
EUR
TCIAI1104-25G
TCIAI1104-100G
Isodecyl acrylate (mixture of branched chain isomers) stabilizzato
Isodecyl acrylate (mixture of branched chain isomers)
Formula:
C₁₃H₂₄O₂ Peso molecolare: 212,33 g/mol Punto di ebollizione: 158°C Flash Pt: 116°C |
MDL Number:
MFCD00080455 CAS Number: 1330-61-6 EINECS: 215-542-5 UN: 3082 ADR: 9,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | many component mixture |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...