Stampa…

Istradefylline ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

I1100-1G I1100-250MG
TCIAI1100-1GEA 209 EUR
TCIAI1100-1G TCIAI1100-250MG
Istradefylline ≥98.0% (by HPLC, titration analysis)
Istradefylline
Formula: C₂₀H₂₄N₄O₄
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00928421
CAS Number: 155270-99-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 191.0 to 195.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR