Stampa…

(R,R)-2,2'-Isopropylidenebis(4-isopropyl-2-oxazoline) ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: (4R,4'R)-2,2'-(Propane-2,2-diyl)bis(4-isopropyl-4,5-dihydrooxazole), (R,R)-2,2-Bis(4-isopropyl-2-oxazolin-2-yl)propane, 2,2-Bis[(4R)-4-isopropyl-2-oxazolin-2-yl]propane

I1089-250MG I1089-1G
TCIAI1089-250MGEA 71 EUR
TCIAI1089-250MG TCIAI1089-1G
(R,R)-2,2'-Isopropylidenebis(4-isopropyl-2-oxazoline) ≥98.0% (by GC)
(R,R)-2,2'-Isopropylidenebis(4-isopropyl-2-oxazoline)
Formula: C₁₅H₂₆N₂O₂
Peso molecolare: 266,39 g/mol
MDL Number: MFCD28038687
CAS Number: 150529-94-5

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR