Isoorientin ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
Homoorientin, Luteolin 6-C-beta-D-Glucoside
I1087-10MG
I1087-50MG
TCIAI1087-10MGEA
205
EUR
TCIAI1087-10MG
TCIAI1087-50MG
Isoorientin ≥98.0% (by HPLC)
Isoorientin
Formula:
C₂₁H₂₀O₁₁ Peso molecolare: 448,38 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00017433 CAS Number: 4261-42-1 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Carbon) | 53.40 to 57.40 % |
Water | max. 5.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...