1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4,4,5,5-tetramethyl-2-[1-(methylethyl)pyrazol-4-yl]-1,3,2-dioxaborolane, 1-(1-Methylethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1-Isopropyl-1H-pyrazol-4-boronic acid, pinacol ester, 1-Isopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Formula:
C₁₂H₂₁BN₂O₂ Peso molecolare: 236,12 g/mol |
MDL Number:
MFCD11867881 CAS Number: 879487-10-2 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 41.0 to 45.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...