2-Metilpropene ∼6% in toluene
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-Methylpropene, Isobutene, Isobutylene
I1082-100ML
I1082-500ML
TCIAI1082-100MLEA
25
EUR
TCIAI1082-100ML
TCIAI1082-500ML
2-Metilpropene ∼6% in toluene
2-Metilpropene
Formula:
C₄H₈ Peso molecolare: 56,11 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00008898 CAS Number: 115-11-7 EINECS: 204-066-3 UN: 1993 ADR: 3,II |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Concentration (HNMR) | 6.0 to 7.0 w/w% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...