Stampa…

IC261 ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: 3-(2,4,6-Ttrimethoxybenzylidene)indolin-2-one, (E)-3-(2,4,6-trimethoxybenzylidene)indolin-2-one, SU-5607

I1077-10MG I1077-50MG
TCIAI1077-10MGEA 76 EUR
TCIAI1077-10MG TCIAI1077-50MG
IC261 ≥98.0% (by HPLC)
IC261
Formula: C₁₈H₁₇NO₄
Peso molecolare: 311,34 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00118156
CAS Number: 186611-52-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 212.0 to 216.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR