Indolo[3,2,1-jk]carbazole ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₈H₁₁N |
MDL Number:
MFCD01851757 CAS Number: 205-95-8 |
|
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 134.0 to 138.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...