Stampa…

(4-Iodophenyl)acetonitrile ≥95.0% (by GC)

Fornitore: TCI
Pericolo

I1058-1G I1058-5G
TCIAI1058-1GEA 41 EUR
TCIAI1058-1G TCIAI1058-5G
(4-Iodophenyl)acetonitrile ≥95.0% (by GC)
(4-Iodophenyl)acetonitrile
Formula: C₈H₆IN
MDL Number: MFCD00060306
CAS Number: 51628-12-7
UN: 3439
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Melting point 52.0 to 56.0 deg-C

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR