1-Isoquinolinol ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Isocarbostyril, 1-Hydroxyisoquinoline
I1054-1G
I1054-5G
TCIAI1054-1GEA
44
EUR
TCIAI1054-1G
TCIAI1054-5G
1-Isoquinolinol ≥98.0% (by GC)
1-Isoquinolinol
Formula:
C₉H₇NO Peso molecolare: 145,16 g/mol |
MDL Number:
MFCD00006899 CAS Number: 491-30-5 EINECS: 207-732-1 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 212.0 to 216.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...