Stampa…

6-Iodo-m-toluidine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Amino-4-iodotoluene, 2-Iodo-5-methylaniline

I1034-1G I1034-5G
TCIAI1034-1GEA 34 EUR
TCIAI1034-1G TCIAI1034-5G
6-Iodo-m-toluidine ≥98.0% (by GC, titration analysis)
6-Iodo-m-toluidine
Formula: C₇H₈IN
Peso molecolare: 233,0496 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00833395
CAS Number: 13194-69-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de2ef14197f33e1229d82496d792a208fcff688b&vid=de2ef14197f33e1229d82496d792a208fcff688b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 35.0 to 39.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR