Stampa…

IR 676 iodide ≥97.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1,1,3-Trimethyl-2-(5-(1,1,3-trimethyl-1,3-dihydro-2H-benzo[e]indol-2-ylidene)penta-1,3-dien-1-yl)-1H-benzo[e]indol-3-ium iodide, 1,1,3-Trimethyl-2-[5-(1,1,3-trimethyl-1,3-dihydro-2H-benzo[e]indol-2-ylidene)-1,3-pentadienyl]-1H-benzo[e]indolium iodide

I1030-200MG I1030-1G
TCIAI1030-200MGEA 82 EUR
TCIAI1030-200MG TCIAI1030-1G
IR 676 iodide ≥97.0% (by HPLC, total nitrogen)
IR 676 iodide
Formula: C₃₅H₃₅IN₂
Peso molecolare: 610,58 g/mol
MDL Number: MFCD07433812
CAS Number: 56289-64-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bfde86cdd530d48d79f6712370761f77a70ab3b6&vid=bfde86cdd530d48d79f6712370761f77a70ab3b6&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Lambda max. 675.0 to 685.0 nm (EtOH)
Absorbance (E1%1cm) min. 3400 (EtOH, 675.0 to 685.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR