Stampa…

4-Iodo-2-nitroaniline ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

I1029-1G I1029-5G
TCIAI1029-1GEA 28 EUR
TCIAI1029-1G TCIAI1029-5G
4-Iodo-2-nitroaniline ≥98.0% (by GC)
4-Iodo-2-nitroaniline
Formula: C₆H₅IN₂O₂
Peso molecolare: 264,02 g/mol
MDL Number: MFCD06808526
CAS Number: 20691-72-9

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 124.0 to 128.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR