Stampa…

3-Iodo-1H-pyrazole ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Iodopyrazole

I1016-1G I1016-5G
TCIAI1016-1GEA 71 EUR
TCIAI1016-1G TCIAI1016-5G
3-Iodo-1H-pyrazole ≥97.0% (by GC)
3-Iodo-1H-pyrazole
Formula: C₃H₃IN₂
Peso molecolare: 193,98 g/mol
MDL Number: MFCD12022324
CAS Number: 4522-35-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f650cd569c3fe8c406865025b46b6d0afded43b&vid=0f650cd569c3fe8c406865025b46b6d0afded43b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 68.0 to 72.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR