2-(2-Isopropyl-5-methylphenoxy)acetic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
(2-Isopropyl-5-methylphenoxy)acetic acid
Formula:
C₁₂H₁₆O₃ Peso molecolare: 208.26 g/mol |
MDL Number:
MFCD00021757 CAS Number: 5333-40-4 |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 149.0 to 153.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...