Stampa…

Isopentyl-4-(dimethylamino)benzoate ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Isoamyl 4-(dimethylamino)benzoate, 4-(Dimethylamino)benzoic acid isopentyl ester

I1013-25G I1013-100G
TCIAI1013-25GEA 39 EUR
TCIAI1013-25G TCIAI1013-100G
Isopentyl-4-(dimethylamino)benzoate ≥98.0% (by GC, titration analysis)
Isopentyl-4-(dimethylamino)benzoate
Formula: C₁₄H₂₁NO₂
Peso molecolare: 235,33 g/mol
MDL Number: MFCD00059356
CAS Number: 21245-01-2
EINECS: 244-288-8
Merck Index: 14,03232
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85ddac62d9777d7be15b4bed6407a560c8dd5270&vid=85ddac62d9777d7be15b4bed6407a560c8dd5270&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 35.0 to 39.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR