Stampa…

2-Imidazolecarboxylic acid ≥98.0% (by HPLC)

Fornitore: TCI
Attenzione

I1008-1G I1008-5G
TCIAI1008-1GEA 58 EUR
TCIAI1008-1G TCIAI1008-5G
2-Imidazolecarboxylic acid ≥98.0% (by HPLC)
2-Imidazolecarboxylic acid
Formula: C₄H₄N₂O₂
Punto di fusione: 164°C
MDL Number: MFCD00270816
CAS Number: 16042-25-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c864c2dc4c52518fe492e22c25fe06ce53f6baa7&vid=c864c2dc4c52518fe492e22c25fe06ce53f6baa7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Dark blue powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR