(3-Iodophenyl)acetonitrile ≥96.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
3-Iodobenzyl cyanide, 2-(3-Iodophenyl)acetonitrile
I1002-1G
I1002-5G
TCIAI1002-1GEA
64
EUR
TCIAI1002-1G
TCIAI1002-5G
(3-Iodophenyl)acetonitrile ≥96.0% (by GC)
(3-Iodophenyl)acetonitrile
Formula:
C₈H₆IN Peso molecolare: 243,05 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00040890 CAS Number: 130723-54-5 UN: 3439 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to lump |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...