1-Isopropylpyrazole ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-Isopropyl-1H-pyrazole
I0999-1ML
I0999-5ML
TCIAI0999-1MLEA
39
EUR
TCIAI0999-1ML
TCIAI0999-5ML
1-Isopropylpyrazole ≥98.0% (by GC, titration analysis)
1-Isopropylpyrazole
Formula:
C₆H₁₀N₂ Peso molecolare: 110,16 g/mol Punto di ebollizione: 142°C |
MDL Number:
MFCD07468920 CAS Number: 18952-87-9 UN: 1993 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...