2-Isopropylbenzoic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
I0997-1G
I0997-5G
TCIAI0997-1GEA
76
EUR
TCIAI0997-1G
TCIAI0997-5G
2-Isopropylbenzoic acid ≥98.0% (by GC, titration analysis)
2-Isopropylbenzoic acid
Formula:
C₁₀H₁₂O₂ Peso molecolare: 164,20 g/mol |
MDL Number:
MFCD00045836 CAS Number: 2438-04-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 62.0 to 66.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...