Stampa…

1H,1H,12H,12H-Eicosafluoro-1,12-dodecandiol ≥96.0% (by GC)

Fornitore: TCI

Synonyms: 1H,1H,12H,12H-Perfluoro-1,12-dodecanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Eicosafluoro-1,12-dodecandiol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-Icosafluorododecane-1,12-diol, 1H,1H,12H,12H-Icosafluoro-1,12-dodecanediol, 1H,1H,12H,12H-Perfluoro-1,12-dodecandiol

I0994-1G I0994-5G
TCIAI0994-1GEA 22 EUR
TCIAI0994-1G TCIAI0994-5G
1H,1H,12H,12H-Eicosafluoro-1,12-dodecandiol ≥96.0% (by GC)
1H,1H,12H,12H-Eicosafluoro-1,12-dodecandiol
Formula: C₁₂H₆F₂₀O₂
Peso molecolare: 562,15 g/mol
MDL Number: MFCD00236625
CAS Number: 183162-43-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01b2574e51ea2cd9f589b07323fd21c431952aef&vid=01b2574e51ea2cd9f589b07323fd21c431952aef&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 165.0 to 169.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR