Stampa…

2-Iodo-5-nitrotoluene ≥96.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-Iodo-2-methyl-4-nitrobenzene

I0992-5G I0992-25G I0992-1G
TCIAI0992-5GEA 124 EUR
TCIAI0992-5G TCIAI0992-25G TCIAI0992-1G
2-Iodo-5-nitrotoluene ≥96.0% (by GC)
2-Iodo-5-nitrotoluene
Formula: C₇H₆INO₂
Peso molecolare: 263,03 g/mol
MDL Number: MFCD00051517
CAS Number: 5326-38-5
EINECS: 226-204-1

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 96.0 %
Melting point 100.0 to 104.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR