Stampa…

2-(1H-Imidazol-1-yl)acetonitrile ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 1-(Cyanomethyl)imidazole, (1-Imidazolyl)acetonitrile

I0990-1G I0990-5G
TCIAI0990-1GEA 117 EUR
TCIAI0990-1G TCIAI0990-5G
2-(1H-Imidazol-1-yl)acetonitrile ≥98.0% (by GC, titration analysis)
2-(1H-Imidazol-1-yl)acetonitrile
Formula: C₅H₅N₃
Peso molecolare: 107,12 g/mol
MDL Number: MFCD06421433
CAS Number: 98873-55-3
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77049a24b7b68769f439a7cf0433b9ab301b0cb0&vid=77049a24b7b68769f439a7cf0433b9ab301b0cb0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 54.0 to 58.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR