Stampa…

3-Isopropoxyphenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Isopropoxybenzeneboronic acid

I0984-1G I0984-5G
TCIAI0984-1GEA 33 EUR
TCIAI0984-1G TCIAI0984-5G
3-Isopropoxyphenylboronic acid (contains varying amounts of Anhydride)
3-Isopropoxyphenylboronic acid
Formula: C₉H₁₃BO₃
Peso molecolare: 180,01 g/mol
MDL Number: MFCD03701512
CAS Number: 216485-86-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32845bb21b31f8cd46dbf9fc89e9073d93d288a5&vid=32845bb21b31f8cd46dbf9fc89e9073d93d288a5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 111.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR