Isothiazole ≥98.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Formula:
C₃H₃NS Punto di ebollizione: 114°C |
MDL Number:
MFCD00020818 CAS Number: 288-16-4 UN: 1993 ADR: 3,II |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...