2-Iodobiphenyl ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
2-Iodo-1,1'-biphenyl
I0967-5G
I0967-25G
TCIAI0967-5GEA
40
EUR
TCIAI0967-5G
TCIAI0967-25G
2-Iodobiphenyl ≥98.0% (by GC)
2-Iodobiphenyl
Formula:
C₁₂H₉I Peso molecolare: 280,11 g/mol Punto di ebollizione: 158°C |
MDL Number:
MFCD00039396 CAS Number: 2113-51-1 EINECS: 218-303-3 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...